missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrin beta 1 Antibody (4C3), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB300-574-0.025ml
This item is not returnable.
View return policy
Description
Spectrin beta 1 Monoclonal specifically detects Spectrin beta 1 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Spectrin beta 1 | |
| Monoclonal | |
| Unconjugated | |
| beta-I spectrin, EL3, erythrocyte, HS2, HSPTB1, membrane cytoskeletal protein, spectrin, beta, erythrocytic, SPTB1 | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| Detects spectrin from erythrocytes, brain and muscle cells. This has been shown to specifically detect the two known alternatively spliced forms of spectrin, beta-1 epsilon-1, present in erythrocytes, and beta-1 epsilon-2, present in nerve and striated muscle cells. It does not cross-react with alpha-2 spectrin or either of the fodrin subunits. | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| 4C3 | |
| Western Blot 1:100, Flow Cytometry 0.5 - 2 μg, Immunohistochemistry 1:20 - 1:200, Immunocytochemistry/Immunofluorescence 1:10 - 1:200, Immunohistochemistry-Paraffin 1:20 - 1:200, Immunohistochemistry-Frozen 1:20 - 1:200 | |
| SPTB | |
| Purified human erythrocyte beta-1 spectrin. | |
| 0.025 mL | |
| Cytoskeleton Markers | |
| 6710 | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction