missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPATA18 Antibody - BSA Free, Novus Biologicals™
Chicken Polyclonal Antibody
178.00€ - 507.00€
Specifications
| Antigen | SPATA18 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18647425
|
Novus Biologicals
NBP2-41148-0.025mg |
0.025 mg |
178.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18137867
|
Novus Biologicals
NBP2-41148 |
0.1 mg |
507.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SPATA18 Polyclonal specifically detects SPATA18 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
| SPATA18 | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 132671 | |
| Antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human SPATA18. The immunogen is located within amino acids 90 - 140 of SPATA18. Amino Acid Squence: CDTFDRERHKDPSPRDRDMQ | |
| Primary | |
| At least three isoforms of SPATA18 are known to exist; this antibody will detect the two longest isoforms. SPATA18 antibody is predicted to not cross-react with other SPATA family members. |
| 1 mg/mL | |
| Polyclonal | |
| Chicken | |
| Human, Mouse, Rat | |
| FLJ32906, Mieap, mitochondria-eating protein, spermatogenesis associated 18 homolog (rat), Spermatogenesis-associated protein 18, SPETEX1 | |
| SPATA18 | |
| IgY | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title