missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPATA18 Antibody - BSA Free, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP2-41148
This item is not returnable.
View return policy
Description
SPATA18 Polyclonal specifically detects SPATA18 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| SPATA18 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| SPATA18 | |
| Antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human SPATA18. The immunogen is located within amino acids 90 - 140 of SPATA18. Amino Acid Squence: CDTFDRERHKDPSPRDRDMQ | |
| 0.1 mg | |
| Primary | |
| At least three isoforms of SPATA18 are known to exist; this antibody will detect the two longest isoforms. SPATA18 antibody is predicted to not cross-react with other SPATA family members. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot 1 ug/ml, ELISA | |
| FLJ32906, Mieap, mitochondria-eating protein, spermatogenesis associated 18 homolog (rat), Spermatogenesis-associated protein 18, SPETEX1 | |
| Chicken | |
| Affinity Purified | |
| RUO | |
| 132671 | |
| Human, Mouse, Rat | |
| IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction