missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX4 Antibody (CL5634), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61420-25ul
This item is not returnable.
View return policy
Description
SOX4 Monoclonal specifically detects SOX4 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SOX4 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2), containing 40% glycerol with 0.02% Sodium Azide | |
| SOX4 | |
| This antibody was developed against a synthetic peptide corresponding to amino acids: AGADPSDPLGLYEEEGAG | |
| 25 μL | |
| Stem Cell Markers, Stem Cells | |
| 6659 | |
| Human, Mouse, Rat | |
| Purified |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| CL5634 | |
| Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| ecotropic viral integration site 16, EVI16, SRY (sex determining region Y)-box 4, SRY-related HMG-box gene 4, transcription factor SOX-4 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Specificity of human SOX4 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction