missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX4 Antibody (CL5634), Novus Biologicals™
Mouse Monoclonal Antibody
369.00€ - 539.00€
Specifications
| Antigen | SOX4 |
|---|---|
| Clone | CL5634 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18205703
|
Novus Biologicals
NBP2-61420 |
100 μL |
539.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18697197
|
Novus Biologicals
NBP2-61420-25ul |
25 μL |
369.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SOX4 Monoclonal specifically detects SOX4 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| SOX4 | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| 6659 | |
| This antibody was developed against a synthetic peptide corresponding to amino acids: AGADPSDPLGLYEEEGAG | |
| Primary | |
| Specificity of human SOX4 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| CL5634 | |
| Monoclonal | |
| Purified | |
| Stem Cell Markers, Stem Cells | |
| ecotropic viral integration site 16, EVI16, SRY (sex determining region Y)-box 4, SRY-related HMG-box gene 4, transcription factor SOX-4 | |
| SOX4 | |
| IgG2b | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title