Learn More
OCIL/CLEC2d Antibody (166C1309), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand:  Novus Biologicals NB100-56553
 View more versions of this product
Product Code. 18096775
  • 486.00€ / 0.10mg
Description
OCIL/CLEC2d Monoclonal specifically detects OCIL/CLEC2d in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OCIL/CLEC2d
Monoclonal
0.5 mg/mL
Western Blot
CLAXLLT-1, C-type lectin domain family 2 member D, C-type lectin domain family 2, member D, C-type lectin related f, Lectin-like NK cell receptor, Lectin-like transcript 1, LLT1Osteoclast inhibitory lectin, OCILmember D
Mouse
Protein G purified
RUO
Primary
Mouse
Purified
Western Blot
166C1309
Unconjugated
AAK50881
CLEC2D
This antibody was developed by immunizing mice with a synthetic peptide containing amino acids 2-18 (CVTKASLPMLSPTGSPQ) of mouse OCIL, GenBank no. gb AAL37198.1.
0.1 mg
Immunology
29121
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.