missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCIL/CLEC2d Antibody (166C1309), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
213.00€ - 486.00€
Specifications
| Antigen | OCIL/CLEC2d |
|---|---|
| Clone | 166C1309 |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18017073
|
Novus Biologicals
NB100-56553SS |
0.025 mg |
213.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18096775
|
Novus Biologicals
NB100-56553 |
0.1 mg |
486.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OCIL/CLEC2d Monoclonal specifically detects OCIL/CLEC2d in Mouse samples. It is validated for Western Blot.Specifications
| OCIL/CLEC2d | |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Immunology | |
| AAK50881 | |
| 29121 | |
| This antibody was developed by immunizing mice with a synthetic peptide containing amino acids 2-18 (CVTKASLPMLSPTGSPQ) of mouse OCIL, GenBank no. gb AAL37198.1. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 166C1309 | |
| Western Blot | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| CLAXLLT-1, C-type lectin domain family 2 member D, C-type lectin domain family 2, member D, C-type lectin related f, Lectin-like NK cell receptor, Lectin-like transcript 1, LLT1Osteoclast inhibitory lectin, OCILmember D | |
| CLEC2D | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title