missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX18 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
168.00€ - 507.00€
Specifications
| Antigen | SOX18 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18630489
|
Novus Biologicals
NBP2-41289-0.025mg |
0.025 mg |
168.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18108118
|
Novus Biologicals
NBP2-41289 |
0.1 mg |
507.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SOX18 Polyclonal specifically detects SOX18 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
| SOX18 | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 54345 | |
| Antibody was raised against a 17 amino acid peptide near the amino terminus of human SOX18. The immunogen is located within amino acids 40 - 90 of SOX18. Amino Acid Squence: ASPPSPQRSPPRSPEPG | |
| Primary | |
| SOX18 is predicted to not cross-react with other SOX proteins. |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| HLTS, SRY (sex determining region Y)-box 18, SRY-box 18, transcription factor SOX-18 | |
| SOX18 | |
| IgG | |
| Affinity Purified | |
| 44 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title