missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX18 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41289
This item is not returnable.
View return policy
Description
SOX18 Polyclonal specifically detects SOX18 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| SOX18 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| SOX18 | |
| Antibody was raised against a 17 amino acid peptide near the amino terminus of human SOX18. The immunogen is located within amino acids 40 - 90 of SOX18. Amino Acid Squence: ASPPSPQRSPPRSPEPG | |
| Affinity Purified | |
| RUO | |
| 54345 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA | |
| HLTS, SRY (sex determining region Y)-box 18, SRY-box 18, transcription factor SOX-18 | |
| Rabbit | |
| 44 kDa | |
| 0.1 mg | |
| Primary | |
| SOX18 is predicted to not cross-react with other SOX proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction