missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX11 Antibody (CL0142), Novus Biologicals™
Mouse Monoclonal Antibody
285.00€ - 564.00€
Specifications
| Antigen | SOX11 |
|---|---|
| Clone | CL0142 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18499911
|
Novus Biologicals
NBP2-14782-25ul |
25ul |
285.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18098353
|
Novus Biologicals
NBP2-14782 |
0.1 mL |
564.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SOX11 Monoclonal specifically detects SOX11 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| SOX11 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| 6664 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: FMVWSKIERRKIMEQSPDMHNAEISKRLGKRWKMLKDSEKIPFIREAERLRLKHMADYPDYKYRPRKKPKMDPSAKPSASQSPEKSAAGGGGGSAGGGAGGAKTSKGSSKK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| CL0142 | |
| Monoclonal | |
| Purified | |
| RUO | |
| SRY (sex determining region Y)-box 11, SRY (sex-determining region Y)-box 11, SRY-related HMG-box gene 11, transcription factor SOX-11 | |
| SOX11 | |
| IgG2a | |
| Protein A purified | |
| Specificity of human SOX11 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title