missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX11 Antibody (CL0142), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-14782
This item is not returnable.
View return policy
Description
SOX11 Monoclonal specifically detects SOX11 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SOX11 | |
| Monoclonal | |
| Unconjugated | |
| SRY (sex determining region Y)-box 11, SRY (sex-determining region Y)-box 11, SRY-related HMG-box gene 11, transcription factor SOX-11 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 6664 | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| CL0142 | |
| Western Blot 1 μg/mL, Simple Western 5 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:200-1:500 | |
| SOX11 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: FMVWSKIERRKIMEQSPDMHNAEISKRLGKRWKMLKDSEKIPFIREAERLRLKHMADYPDYKYRPRKKPKMDPSAKPSASQSPEKSAAGGGGGSAGGGAGGAKTSKGSSKK | |
| 0.1 mL | |
| Primary | |
| Specificity of human SOX11 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction