missing translation for 'onlineSavingsMsg'
Learn More

SNIP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

216.00€ - 326.00€

Specifications

Antigen SNIP1
Dilution Western Blot 1:500-1:2000, Immunoprecipitation 1-5 μL/mg of lysate
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18627862
View Documents
Novus Biologicals
NBP2-98468-100ul
100 μg
326.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18677372
View Documents
Novus Biologicals
NBP2-98468-50ul
50 μg
216.00€
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SNIP1 Polyclonal antibody specifically detects SNIP1 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications

Specifications

SNIP1
Western Blot, Immunoprecipitation
Unconjugated
Rabbit
Cell Biology, Signal Transduction
PBS (pH 7.0)
79753
IgG
Antigen and protein A Affinity-purified
Western Blot 1:500-1:2000, Immunoprecipitation 1-5 μL/mg of lysate
Polyclonal
Purified
RUO
Human
dJ423B22.2, FHA domain-containing protein SNIP1, FLJ12553, RP3-423B22.3, Smad nuclear interacting protein (SNIP1), Smad nuclear interacting protein 1, smad nuclear-interacting protein 1
Produced in rabbits immunized with E. coli-derived Human SNIP1 fragment.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.