missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNIP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98468-100ul
This item is not returnable.
View return policy
Description
SNIP1 Polyclonal antibody specifically detects SNIP1 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| SNIP1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-5 μL/mg of lysate | |
| dJ423B22.2, FHA domain-containing protein SNIP1, FLJ12553, RP3-423B22.3, Smad nuclear interacting protein (SNIP1), Smad nuclear interacting protein 1, smad nuclear-interacting protein 1 | |
| Produced in rabbits immunized with E. coli-derived Human SNIP1 fragment. | |
| 100 μg | |
| Cell Biology, Signal Transduction | |
| 79753 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction