missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smith Antigen Antibody (Y12), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
190.00€
Specifications
| Antigen | Smith Antigen |
|---|---|
| Clone | Y12 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 0.5 - 1.0 μg/mL, ELISA, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50 - 1:100, Immunomicroscopy 1:10 - 1:500 |
| Applications | Western Blot |
Description
Smith Antigen Monoclonal specifically detects Smith Antigen in Human, Mouse, Rat, Porcine, Drosophila, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunomicroscopy.Specifications
| Smith Antigen | |
| 0.2 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| The immunogen is generated from human Smith Antigen. | |
| Primary | |
| Store at 4°C. Do not freeze. |
| Y12 | |
| Western Blot 0.5 - 1.0 μg/mL, ELISA, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50 - 1:100, Immunomicroscopy 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Smith antigen | |
| IgG3 κ | |
| Protein G purified | |
| Ab-1 recognizes cross-reactive epitopes on the B'/B and D polypeptides of Sm. Ab-1 precipitates the small nuclear RNAs U1, U2, U4, U5, and U6 providing direct evidence that the Sm antigen resides on all these RNA-protein complexes. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title