missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smith Antigen Antibody (Y12), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NB600-546-100ul
This item is not returnable.
View return policy
Description
Smith Antigen Monoclonal specifically detects Smith Antigen in Human, Mouse, Rat, Porcine, Drosophila, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunomicroscopy.
Specifications
| Smith Antigen | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 0.5 - 1.0 μg/mL, ELISA, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50 - 1:100, Immunomicroscopy 1:10 - 1:500 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Ab-1 recognizes cross-reactive epitopes on the B'/B and D polypeptides of Sm. Ab-1 precipitates the small nuclear RNAs U1, U2, U4, U5, and U6 providing direct evidence that the Sm antigen resides on all these RNA-protein complexes. | |
| Store at 4°C. Do not freeze. | |
| IgG3 κ |
| Western Blot | |
| Y12 | |
| Unconjugated | |
| Smith antigen | |
| The immunogen is generated from human Smith Antigen. | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction