missing translation for 'onlineSavingsMsg'
Learn More

Smad4 Mouse anti-Human, Clone: SMAD4/2440, Novus Biologicals™

Mouse Monoclonal Antibody

508.00€

Specifications

Antigen Smad4
Clone SMAD4/2440
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array
Applications ELISA, Peptide Array
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18697306
View Documents
Novus Biologicals
NBP3-07262-100ug
100 μg
508.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Smad4 Monoclonal specifically detects Smad4 in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.
Specifications

Specifications

Smad4
0.2 mg/mL
ELISA, Peptide Array
Unconjugated
Mouse
Signal Transduction, Stem Cell Signaling Pathway
10 mM PBS with 0.05% BSA
4089
IgG2a κ
Protein A or G purified
SMAD4/2440
Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array
Monoclonal
Purified
RUO
Human
DPC4SMAD, mothers against DPP homolog 4 (Drosophila), hSMAD4, JIP, MAD homolog 4, MADH4mothers against decapentaplegic homolog 4, mothers against decapentaplegic homolog 4, mothers against decapentaplegic, Drosophila, homolog of, 4, SMAD 4, SMAD family member 4MAD, mothers against decapentaplegic homolog 4 (Drosophila), SMAD, mothers against DPP homolog 4, Smad4
Recombinant full-length human Smad4 protein
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.