missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smad4 Mouse anti-Human, Clone: SMAD4/2440, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07262-100ug
This item is not returnable.
View return policy
Description
Smad4 Monoclonal specifically detects Smad4 in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.
Specifications
| Smad4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| DPC4SMAD, mothers against DPP homolog 4 (Drosophila), hSMAD4, JIP, MAD homolog 4, MADH4mothers against decapentaplegic homolog 4, mothers against decapentaplegic homolog 4, mothers against decapentaplegic, Drosophila, homolog of, 4, SMAD 4, SMAD family member 4MAD, mothers against decapentaplegic homolog 4 (Drosophila), SMAD, mothers against DPP homolog 4, Smad4 | |
| Recombinant full-length human Smad4 protein | |
| 100 μg | |
| Signal Transduction, Stem Cell Signaling Pathway | |
| 4089 | |
| Store at 4°C. | |
| IgG2a κ |
| ELISA, Peptide Array | |
| SMAD4/2440 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction