Learn More
Zeiss™ Slider
Use these sliders for a variety of applications
51.00€ - 1531.70€
Product Code | Brand | Type | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Description | Price | Quantity & Availability | ||||
11835093
|
Zeiss™
426968-9030-000 |
Slider | 50x/0.7 Epi+ |
1526.40€
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
12312043
|
Zeiss™
426717-9020-000 |
Slider with Fixed Phase Stop Ph 1/0.4 | with fixed phase stop Ph 1/0.4 |
51.00€
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
11825093
|
Zeiss™
426968-9020-000 |
Slider | 5x/0.13, 10x/0.2, 20x/0.4 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
Description
Slider 10x46 mm Ph/PlasDIC, H, Ph/PlasDIC- 2 positions for Ph or PlasDIC diaphragms
- brightfield position with neutral
- density filter 0.06
- With fixed phase stop Ph 1/0.4
- 5x/0.13, 10x/0.2, 20x/0.4 Epi+
- 50x/0.7 Epi+
Specifications
1 Ea. |