Learn More
Zeiss™ Slider
Use these sliders for a variety of applications
Brand: Zeiss™ 426968-9030-000
Additional Details : Weight : 0.01000kg
Description
Slider 10x46 mm Ph/PlasDIC, H, Ph/PlasDIC- 2 positions for Ph or PlasDIC diaphragms
- brightfield position with neutral
- density filter 0.06
- With fixed phase stop Ph 1/0.4
- 5x/0.13, 10x/0.2, 20x/0.4 Epi+
- 50x/0.7 Epi+
Specifications
Slider | |
50x/0.7 Epi+ | |
1 Ea. |