Learn More
Zeiss™ Slider
Click to view available options
Type:
Slider
Slider with Fixed Phase Stop Ph 1/0.4
Description:
DIC slider EC EP
DIC slider EC EP Epi+
Slider w/ Fixed Phase Stop Ph 1/0.4
Unit Size:
Each
Description
Slider 10x46 mm Ph/PlasDIC, H, Ph/PlasDIC
- 2 positions for Ph or PlasDIC diaphragms
- brightfield position with neutral
- density filter 0.06
- With fixed phase stop Ph 1/0.4
- 5x/0.13, 10x/0.2, 20x/0.4 Epi+
- 50x/0.7 Epi+
Specifications
Specifications
| Type | Slider |
| Description | DIC slider EC EP Epi+ |
| Quantity | 1 Ea. |