missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC39A11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-58224
This item is not returnable.
View return policy
Description
SLC39A11 Polyclonal specifically detects SLC39A11 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| SLC39A11 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| C17orf26, chromosome 17 open reading frame 26, solute carrier family 39 (metal ion transporter), member 11, Solute carrier family 39 member 11, zinc transporter ZIP11, ZIP11, ZIP-11, Zrt- and Irt-like protein 11 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 201266 | |
| Human | |
| IgG |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| SLC39A11 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TASATFESARNLAIGIGIQNFPEGLAVSLPLRGAGFSTWRAFW | |
| 100 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction