missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCAM/CD146 Antibody (6C3E6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37365-0.025ml
This item is not returnable.
View return policy
Description
MCAM/CD146 Monoclonal specifically detects MCAM/CD146 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications
| MCAM/CD146 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready | |
| CD146, CD146 antigen, cell surface glycoprotein MUC18, Cell surface glycoprotein P1H12, melanoma adhesion molecule, melanoma cell adhesion moleculeS-endo 1 endothelial-associated antigen, Melanoma-associated antigen A32, Melanoma-associated antigen MUC18, MUC18Gicerin | |
| Mouse | |
| 71.6 kDa | |
| 0.025 mL | |
| Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 4162 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| 6C3E6 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| MCAM | |
| Purified recombinant fragment of human MCAM (AA: 84-189) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction