missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCAM/CD146 Antibody (6C3E6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
217.00€ - 557.00€
Specifications
| Antigen | MCAM/CD146 |
|---|---|
| Clone | 6C3E6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18162828
|
Novus Biologicals
NBP2-37365 |
0.1 mL |
557.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18690656
|
Novus Biologicals
NBP2-37365-0.025ml |
0.025 mL |
217.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MCAM/CD146 Monoclonal specifically detects MCAM/CD146 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| MCAM/CD146 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| RUO | |
| Human | |
| CD146, CD146 antigen, cell surface glycoprotein MUC18, Cell surface glycoprotein P1H12, melanoma adhesion molecule, melanoma cell adhesion moleculeS-endo 1 endothelial-associated antigen, Melanoma-associated antigen A32, Melanoma-associated antigen MUC18, MUC18Gicerin | |
| MCAM | |
| IgG1 | |
| Affinity Purified |
| 6C3E6 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| PBS with 0.05% Sodium Azide | |
| 4162 | |
| Purified recombinant fragment of human MCAM (AA: 84-189) expressed in E. Coli. | |
| Primary | |
| 71.6 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title