missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Skp1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
202.00€ - 573.00€
Specifications
| Antigen | Skp1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30227217
|
Novus Biologicals
NBP3-35243-20ul |
20 μL |
202.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30232261
|
Novus Biologicals
NBP3-35243-100ul |
100 μL |
573.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Skp1 Polyclonal antibody specifically detects Skp1 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| Skp1 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Wnt Signaling Pathway | |
| PBS (pH 7.3), 50% glycerol | |
| 6500 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Cyclin-A/CDK2-associated protein p19, EMC19p19skp1, MGC34403, OCP2OCP-2, OCP-IIS-phase kinase-associated protein 1A (p19A), Organ of Corti protein 2, Organ of Corti protein II, p19Acyclin A/CDK2-associated p19, RNA polymerase II elongation factor-like protein, RNA polymerase II elongation factor-like protein OCP2, SKP1Acyclin A/CDK2-associated protein p19, S-phase kinase-associated protein 1, TCEB1LSIII, Transcription elongation factor B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-163 of human Skp1 (NP_733779.1).,, Sequence:, MPSIKLQSSDGEIFEVDVEIAKQSVTIKTMLEDLGMDDEGDDDPVPLPNVNAAILKKVIQWCTHHKDDPPPPEDDENKEKRTDDIPVWDQEFLKVDQGTLFELILAANYLDIKGLLDVTCKTVANMIKGKTPEEIRKTFNIKNDFTEEEEAQVRKENQWCEEK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title