missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Skp1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35243-20ul
This item is not returnable.
View return policy
Description
Skp1 Polyclonal antibody specifically detects Skp1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| Skp1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| Cyclin-A/CDK2-associated protein p19, EMC19p19skp1, MGC34403, OCP2OCP-2, OCP-IIS-phase kinase-associated protein 1A (p19A), Organ of Corti protein 2, Organ of Corti protein II, p19Acyclin A/CDK2-associated p19, RNA polymerase II elongation factor-like protein, RNA polymerase II elongation factor-like protein OCP2, SKP1Acyclin A/CDK2-associated protein p19, S-phase kinase-associated protein 1, TCEB1LSIII, Transcription elongation factor B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-163 of human Skp1 (NP_733779.1).,, Sequence:, MPSIKLQSSDGEIFEVDVEIAKQSVTIKTMLEDLGMDDEGDDDPVPLPNVNAAILKKVIQWCTHHKDDPPPPEDDENKEKRTDDIPVWDQEFLKVDQGTLFELILAANYLDIKGLLDVTCKTVANMIKGKTPEEIRKTFNIKNDFTEEEEAQVRKENQWCEEK | |
| 20 μL | |
| Wnt Signaling Pathway | |
| 6500 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction