missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRT7 Antibody (1E2B2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
203.00€ - 552.00€
Specifications
| Antigen | SIRT7 |
|---|---|
| Clone | 1E2B2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18228880
|
Novus Biologicals
NBP2-52564 |
0.1 mg |
552.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18626517
|
Novus Biologicals
NBP2-52564-0.025mg |
0.025 mg |
203.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SIRT7 Monoclonal specifically detects SIRT7 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
| SIRT7 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| DNA Repair, Epigenetics, Histone Deacetylases | |
| PBS with 0.05% Sodium Azide | |
| 51547 | |
| Purified recombinant fragment of human SIRT7 (AA: 1-105) expressed in E. Coli. | |
| Primary | |
| 44.9 kDa |
| 1E2B2 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.5.1.-, MGC126840, MGC126842, silent mating type information regulation 2, S.cerevisiae, homolog 7, SIR2L7NAD-dependent deacetylase sirtuin-7, SIR2-like protein 7, sir2-related protein type 7, sirtuin (silent mating type information regulation 2 homolog) 7 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S.cerevisiae, homolog) 7, sirtuin 7, sirtuin type 7 | |
| SIRT7 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title