missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRT7 Antibody (1E2B2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52564-0.025mg
This item is not returnable.
View return policy
Description
SIRT7 Monoclonal specifically detects SIRT7 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications
| SIRT7 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready | |
| EC 3.5.1.-, MGC126840, MGC126842, silent mating type information regulation 2, S.cerevisiae, homolog 7, SIR2L7NAD-dependent deacetylase sirtuin-7, SIR2-like protein 7, sir2-related protein type 7, sirtuin (silent mating type information regulation 2 homolog) 7 (S. cerevisiae), sirtuin (silent mating type information regulation 2, S.cerevisiae, homolog) 7, sirtuin 7, sirtuin type 7 | |
| Mouse | |
| 44.9 kDa | |
| 0.025 mg | |
| DNA Repair, Epigenetics, Histone Deacetylases | |
| 51547 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry | |
| 1E2B2 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| SIRT7 | |
| Purified recombinant fragment of human SIRT7 (AA: 1-105) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction