missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SIAH1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00€ - 508.00€
Specifications
| Antigen | SIAH1 |
|---|---|
| Dilution | Western Blot : 1-5 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067169
|
Novus Biologicals™
NBP3-30880-100ug |
100 μg |
508.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066922
|
Novus Biologicals™
NBP3-30880-25ug |
25 μg |
213.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SIAH1 Polyclonal antibody specifically detects SIAH1 in Human samples. It is validated for Western BlotSpecifications
| SIAH1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Cell Cycle and Replication, Wnt Signaling Pathway | |
| Phosphate buffered saline | |
| 6477 | |
| IgG | |
| Ammonium sulfate precipitation |
| Western Blot : 1-5 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| E3 ubiquitin-protein ligase SIAH1, EC 6.3.2, EC 6.3.2.-, FLJ08065, hSIAH1, HUMSIAH, Seven in absentia homolog 1, seven in absentia homolog 1 (Drosophila), Siah-1, siah-1a, SIAH1A | |
| Synthetic peptide derived from the human SIAH1 protein. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title