missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SIAH1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30880-100ug
This item is not returnable.
View return policy
Description
SIAH1 Polyclonal antibody specifically detects SIAH1 in Human samples. It is validated for Western Blot
Specifications
| SIAH1 | |
| Polyclonal | |
| Western Blot : 1-5 μg/mL | |
| E3 ubiquitin-protein ligase SIAH1, EC 6.3.2, EC 6.3.2.-, FLJ08065, hSIAH1, HUMSIAH, Seven in absentia homolog 1, seven in absentia homolog 1 (Drosophila), Siah-1, siah-1a, SIAH1A | |
| Synthetic peptide derived from the human SIAH1 protein. | |
| 100 μg | |
| Apoptosis, Cancer, Cell Cycle and Replication, Wnt Signaling Pathway | |
| 6477 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction