missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGLT2/SLC5A2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-92384-25ul
This item is not returnable.
View return policy
Description
SGLT2/SLC5A2 Polyclonal antibody specifically detects SGLT2/SLC5A2 in Human, Mouse, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications
| SGLT2/SLC5A2 | |
| Polyclonal | |
| Western Blot reactivity reported in scientific literature (PMID: 25894829)., Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence reactivity reported in scientific literature (PMID: 25894829)., Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Low affinity sodium-glucose cotransporter, Na(+)/glucose cotransporter 2, SGLT2sodium/glucose cotransporter 2, solute carrier family 5 (sodium/glucose cotransporter), member 2, solute carrier family 5 (sodium/glucose transporter), member 2, Solute carrier family 5 member 2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| SLC5A2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:FHEVGGYSGLFDKYLGAATSLTVSEDPAVGNISSFCYRPRPDSYHLL | |
| 25 μL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 6524 | |
| Human, Mouse, Canine | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction