missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFTS Virus HB29 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
189.00€ - 557.00€
Specifications
| Antigen | SFTS Virus HB29 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18623259
|
Novus Biologicals
NBP2-41156-0.025mg |
0.025 mg |
189.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18110828
|
Novus Biologicals
NBP2-41156 |
0.1 mg |
557.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SFTS Virus HB29 Polyclonal specifically detects SFTS Virus HB29 in Virus samples. It is validated for Western Blot, ELISA.Specifications
| SFTS Virus HB29 | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human SFTS Virus HB29 Membrane Glycoprotein. The immunogen is located within the last 50 amino acids of SFTS Virus HB29. Amino Acid Squence: KLGTKQVFRSRTKLA | |
| Primary | |
| SFTS Virus HB29 Membrane Glycoprotein antibody is specific to the SFTS Virus HB29. |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Virus | |
| B303_sMgp1, Membrane glycoprotein polyprotein | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title