missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFTS Virus HB29 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP2-41156
This item is not returnable.
View return policy
Description
SFTS Virus HB29 Polyclonal specifically detects SFTS Virus HB29 in Virus samples. It is validated for Western Blot, ELISA.
Specifications
| SFTS Virus HB29 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| SFTS Virus HB29 Membrane Glycoprotein antibody is specific to the SFTS Virus HB29. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot, ELISA 1 μg/mL | |
| B303_sMgp1, Membrane glycoprotein polyprotein | |
| Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human SFTS Virus HB29 Membrane Glycoprotein. The immunogen is located within the last 50 amino acids of SFTS Virus HB29. Amino Acid Squence: KLGTKQVFRSRTKLA | |
| 0.1 mg | |
| Primary | |
| Virus | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction