missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpina3g Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | Serpina3g |
|---|---|
| Concentration | 450 μg/mL |
| Applications | Immunohistochemistry, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30233676
|
Proteintech
55353-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30237978
|
Proteintech
55353-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Serpina3g antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of mouse Serpina3g. This antibody recognizes mouse antigen. The Serpina3g antibody has been validated for the following applications: IHC, ELISA analysis.Specifications
| Serpina3g | |
| Immunohistochemistry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| Peptide | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 450 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 20715 | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title