missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpina3g Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55353-1-AP-150UL
This item is not returnable.
View return policy
Description
The Serpina3g antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of mouse Serpina3g. This antibody recognizes mouse antigen. The Serpina3g antibody has been validated for the following applications: IHC, ELISA analysis.Specifications
| Serpina3g | |
| Polyclonal | |
| Unconjugated | |
| Peptide | |
| 150 μL | |
| Primary | |
| Mouse | |
| Polyclonal Antibody | |
| IgG |
| Immunohistochemistry, ELISA | |
| 450 μg/mL | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 20715 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?