missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin B8/Proteinase Inhibitor 8 Antibody (3G2E1D9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
168.00€
Specifications
| Antigen | Serpin B8/Proteinase Inhibitor 8 |
|---|---|
| Clone | 3G2E1D9 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000 |
| Applications | Western Blot |
Description
Serpin B8/Proteinase Inhibitor 8 Monoclonal specifically detects Serpin B8/Proteinase Inhibitor 8 in Human samples. It is validated for Western Blot, ELISA.Specifications
| Serpin B8/Proteinase Inhibitor 8 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| CAP-2, CAP2clade B (ovalbumin), member 8, Cytoplasmic antiproteinase 2, Peptidase inhibitor 8, PI-8, protease inhibitor 8 (ovalbumin type), serpin B8, serpin peptidase inhibitor, clade B (ovalbumin), member 8 | |
| SERPINB8 | |
| IgG1 | |
| Affinity Purified |
| 3G2E1D9 | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Mouse | |
| Cardiovascular Biology | |
| P50452 | |
| 5271 | |
| Ovalbumin | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title