missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin B8/Proteinase Inhibitor 8 Antibody (3G2E1D9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-60510-0.025ml
This item is not returnable.
View return policy
Description
Serpin B8/Proteinase Inhibitor 8 Monoclonal specifically detects Serpin B8/Proteinase Inhibitor 8 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Serpin B8/Proteinase Inhibitor 8 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| CAP-2, CAP2clade B (ovalbumin), member 8, Cytoplasmic antiproteinase 2, Peptidase inhibitor 8, PI-8, protease inhibitor 8 (ovalbumin type), serpin B8, serpin peptidase inhibitor, clade B (ovalbumin), member 8 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot | |
| 3G2E1D9 | |
| Unconjugated | |
| P50452 | |
| SERPINB8 | |
| Ovalbumin | |
| 0.025 mL | |
| Cardiovascular Biology | |
| 5271 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction