missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin A7/TBG Antibody (1C3H11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37323-0.025ml
This item is not returnable.
View return policy
Description
Serpin A7/TBG Monoclonal specifically detects Serpin A7/TBG in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Serpin A7/TBG | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| antitrypsin), member 7, FLJ17820, member 7, Serpin A7, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), T4-binding globulin, TBGclade A (alpha-1 antiproteinase, thyroxin-binding globulin, thyroxine-binding globulin | |
| Mouse | |
| 46.3 kDa | |
| 0.025 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, ELISA | |
| 1C3H11 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| SERPINA7 | |
| Purified recombinant fragment of human SERPINA7 (AA: 168-302) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| 6906 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction