missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin A7/TBG Antibody (1C3H11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
217.00€ - 590.00€
Specifications
| Antigen | Serpin A7/TBG |
|---|---|
| Clone | 1C3H11 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18638605
|
Novus Biologicals
NBP2-37323-0.025ml |
0.025 mL |
217.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18152719
|
Novus Biologicals
NBP2-37323 |
0.1 mL |
590.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Serpin A7/TBG Monoclonal specifically detects Serpin A7/TBG in Human samples. It is validated for Western Blot, ELISA.Spécification
| Serpin A7/TBG | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| 6906 | |
| Purified recombinant fragment of human SERPINA7 (AA: 168-302) expressed in E. Coli. | |
| Primary | |
| 46.3 kDa |
| 1C3H11 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Mouse | |
| Human | |
| antitrypsin), member 7, FLJ17820, member 7, Serpin A7, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), T4-binding globulin, TBGclade A (alpha-1 antiproteinase, thyroxin-binding globulin, thyroxine-binding globulin | |
| SERPINA7 | |
| IgG1 | |
| Affinity Purified |
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit