missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serine Palmitoyltransferase 2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76573-0.025mg
This item is not returnable.
View return policy
Description
Serine Palmitoyltransferase 2 Polyclonal antibody specifically detects Serine Palmitoyltransferase 2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunofluorescence, Immunocytochemistry
Specifications
| Serine Palmitoyltransferase 2 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunofluorescence, Immunocytochemistry | |
| 1 mg/mL | |
| Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 10-20 μg/mL, Immunofluorescence 20 μg/mL, Immunocytochemistry 10 μg/mL | |
| EC 2.3.1, EC 2.3.1.50, KIAA0526HSN1C, LCB 2, LCB2a, LCB2NSAN1C, Long chain base biosynthesis protein 2, Long chain base biosynthesis protein 2a, serine palmitoyltransferase 2, serine palmitoyltransferase, long chain base subunit 2, serine palmitoyltransferase, subunit II, Serine-palmitoyl-CoA transferase 2, SPT 2, SPT2 | |
| Antibody was raised against a peptide corresponding to 15 amino acids near the carboxy terminus of human Serine Palmitoyltransferase 2. The immunogen is located within the last 50 amino acids of Serine Palmitoyltransferase 2. Amino Acid Sequence: LDRPFDETTYEETED | |
| 0.025 mg | |
| Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
| 9517 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction