missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serine Palmitoyltransferase 2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
168.00€ - 507.00€
Specifications
| Antigen | Serine Palmitoyltransferase 2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 10-20 μg/mL, Immunofluorescence 20 μg/mL, Immunocytochemistry 10 μg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunofluorescence, Immunocytochemistry |
| Classification | Polyclonal |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
18598352
|
Novus Biologicals
NBP1-76573-0.025mg |
0.025 mg |
168.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18505574
|
Novus Biologicals
NBP1-76573 |
0.1 mg |
507.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
Serine Palmitoyltransferase 2 Polyclonal antibody specifically detects Serine Palmitoyltransferase 2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunofluorescence, ImmunocytochemistrySpezifikation
| Serine Palmitoyltransferase 2 | |
| Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 10-20 μg/mL, Immunofluorescence 20 μg/mL, Immunocytochemistry 10 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| EC 2.3.1, EC 2.3.1.50, KIAA0526HSN1C, LCB 2, LCB2a, LCB2NSAN1C, Long chain base biosynthesis protein 2, Long chain base biosynthesis protein 2a, serine palmitoyltransferase 2, serine palmitoyltransferase, long chain base subunit 2, serine palmitoyltransferase, subunit II, Serine-palmitoyl-CoA transferase 2, SPT 2, SPT2 | |
| Antibody was raised against a peptide corresponding to 15 amino acids near the carboxy terminus of human Serine Palmitoyltransferase 2. The immunogen is located within the last 50 amino acids of Serine Palmitoyltransferase 2. Amino Acid Sequence: LDRPFDETTYEETED | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
| PBS | |
| 9517 | |
| IgG | |
| Peptide affinity purified |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts