missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S6 Ribosomal protein Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 80208-1-RR-20UL
This item is not returnable.
View return policy
Description
The S6 Ribosomal protein antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human S6 Ribosomal protein. This antibody recognizes human, mouse, rat, zebrafish antigen. The S6 Ribosomal protein antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.Specifications
| S6 Ribosomal protein | |
| Recombinant Monoclonal | |
| 300 μg/mL | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 6194 | |
| Store at -20°C. Stable for one year after shipment. | |
| Recombinant Antibody | |
| IgG |
| Western Blot, Immunocytochemistry/Immunofluorescence, ELISA | |
| 6L9 | |
| Unconjugated | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat, Zebrafish | |
| 32 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction