missing translation for 'onlineSavingsMsg'
Learn More

S6 Ribosomal protein Rabbit Recombinant Antibody, Proteintech

Rabbit Recombinant Antibody

129.00€ - 399.00€

Specifications

Antigen S6 Ribosomal protein
Clone 6L9
Concentration 300 μg/mL
Applications Western Blot, Immunocytochemistry/Immunofluorescence, ELISA
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30236976
View Documents
Proteintech
80208-1-RR-100UL
100 μL
ONLINE EXCLUSIVE PRICE
439.00€
399.00€ / 100µL
Save 40.00€
9% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30235352
View Documents
Proteintech
80208-1-RR-20UL
20 μL
ONLINE EXCLUSIVE PRICE
149.00€
129.00€ / 20µL
Save 20.00€
13% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The S6 Ribosomal protein antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human S6 Ribosomal protein. This antibody recognizes human, mouse, rat, zebrafish antigen. The S6 Ribosomal protein antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.
Specifications

Specifications

S6 Ribosomal protein
300 μg/mL
Recombinant Monoclonal
Liquid
RUO
6194
IgG
Protein A purification
Recombinant Antibody
6L9
Western Blot, Immunocytochemistry/Immunofluorescence, ELISA
Unconjugated
Rabbit
Human, Mouse, Rat, Zebrafish
Peptide
Primary
Store at -20°C. Stable for one year after shipment.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.