missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A1 Antibody (2C8B8), Novus Biologicals™
Mouse Monoclonal Antibody
203.00€
Specifications
| Antigen | S100A1 |
|---|---|
| Clone | 2C8B8 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Western Blot |
Descripción
S100A1 Monoclonal specifically detects S100A1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Especificaciones
| S100A1 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Astrocyte Markers, Breast Cancer, Cell Biology, Cytoskeleton Markers, Neuronal Cell Markers, Neuroscience | |
| Ascites with 0.03% Sodium Azide | |
| protein S100-A1, S100 alpha, S100 calcium binding protein A1, S100 calcium-binding protein A1S100, S-100 protein alpha chain, S-100 protein subunit alpha, S100 protein, alpha polypeptide, S100A, S100-alpha | |
| S100A1 | |
| IgG1 | |
| Unpurified | |
| S100A1 Antibody (2C8B8) |
| 2C8B8 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P23297 | |
| 6271 | |
| Ni-NTA purified truncated recombinant S100A1 Antibody (2C8B8) expressed in E. Coli strain BL21 (DE3) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto