missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A1 Antibody (2C8B8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-60530-0.025ml
This item is not returnable.
View return policy
Description
S100A1 Monoclonal specifically detects S100A1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| S100A1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| P23297 | |
| S100A1 | |
| Ni-NTA purified truncated recombinant S100A1 Antibody (2C8B8) expressed in E. Coli strain BL21 (DE3) | |
| 0.025 mL | |
| Astrocyte Markers, Breast Cancer, Cell Biology, Cytoskeleton Markers, Neuronal Cell Markers, Neuroscience | |
| 6271 | |
| Human | |
| Purified |
| Western Blot | |
| 2C8B8 | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| protein S100-A1, S100 alpha, S100 calcium binding protein A1, S100 calcium-binding protein A1S100, S-100 protein alpha chain, S-100 protein subunit alpha, S100 protein, alpha polypeptide, S100A, S100-alpha | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| S100A1 Antibody (2C8B8) | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction