missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUNX1T1/ETO Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
369.00€
Specifications
| Antigen | RUNX1T1/ETO |
|---|---|
| Dilution | Western Blot 0.04-0.4 μg/mL, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
RUNX1T1/ETO Polyclonal specifically detects RUNX1T1/ETO in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| RUNX1T1/ETO | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 862 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:KGGGSSSSHSRQQSPVNPDPVALDAHREFLHRPASGYVPEEIWKKAE | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.04-0.4 μg/mL, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| AML1T1protein CBFA2T1, CBFA2T1Cyclin-D-related protein, CDRMGC2796, core-binding factor, runt domain, alpha subunit 2; translocated to, 1; cyclinD-related, DKFZp564B213, Eight twenty one protein, ETOFLJ33145, MTG8acute myelogenous leukemia 1 translocation 1, cyclin-D related, Protein ETO, Protein MTG8, runt-related transcription factor 1; translocated to, 1 (cyclin D-related), Zinc finger MYND domain-containing protein 2, ZMYND2myeloid translocation gene on 8q22 | |
| RUNX1T1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title