missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rubicon Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93588-0.1ml
This item is not returnable.
View return policy
Description
Rubicon Polyclonal antibody specifically detects Rubicon in Mouse samples. It is validated for Western Blot
Specifications
| Rubicon | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Baron, Beclin 1-interacting protein, Beclin-1 Associated RUN Domain Containing Protein, KIAA0226, Rubicon, RUN And Cysteine Rich Domain Containing Beclin 1 Interacting Protein, RUN Domain And Cysteine-Rich Domain Containing, Beclin 1-Interacting Protein, Run Domain Beclin-1-Interacting And Cysteine-Rich Domain-Containing Protein, rundataxin, SCAR15 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human Rubicon (NP_055502.1). MRPEGAGMELGGGEERLPEESRREHWQLLGNLKTTVEGLVSTNSPNVWSKYGGLERLCRDMQSILYHGLIRDQACRRQTDYWQFVKDIRWLSPHSALHVEKFISVHENDQSSADGASERAVAELWLQHSLQYHCLSAQLRPLLGDRQYIRKFYTDAAFLLSDAHVTAMLQCLEAVEQNNPRLLAQIDASMFARKHESPLL | |
| 0.1 mL | |
| Autophagy, Cancer, Cardiovascular Biology, Cell Biology, Endocrinology, Signal Transduction | |
| 9711 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction