missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPAP1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
129.00€ - 349.00€
Specifications
| Antigen | RPAP1 |
|---|---|
| Clone | 2A6H5 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA, Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235874
|
Proteintech
68193-1-IG-20UL |
20 μL |
145.00€ 129.00€ / 20µL Save 16.00€ 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234326
|
Proteintech
68193-1-IG-150UL |
150 μL |
410.00€ 349.00€ / 150µL Save 61.00€ 14% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The RPAP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RPAP1. This antibody recognizes human, mouse, rat antigen. The RPAP1 antibody has been validated for the following applications: WB, ELISA, FC (Intra) analysis.Specifications
| RPAP1 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 26015 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 2A6H5 | |
| Western Blot, ELISA, Flow Cytometry (Intracellular Staining) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| DKFZp727M111; FLJ12732; KIAA1403; MGC858 | |
| IgG1 | |
| Protein G purification | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title