missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPAP1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68193-1-IG-20UL
This item is not returnable.
View return policy
Description
The RPAP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RPAP1. This antibody recognizes human, mouse, rat antigen. The RPAP1 antibody has been validated for the following applications: WB, ELISA, FC (Intra) analysis.Specifications
| RPAP1 | |
| Monoclonal | |
| 1000 μg/mL | |
| DKFZp727M111; FLJ12732; KIAA1403; MGC858 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 153 kDa | |
| Liquid |
| Western Blot, ELISA, Flow Cytometry (Intracellular Staining) | |
| 2A6H5 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 26015 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur