Learn More
Fisherbrand™ Round Glass Narrow Mouth Alpha Bottles
Product Code. p-8009859 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
100 mL
125 mL
250 mL
500 mL
1000 mL
Color:
Amber
Clear
Unit Size:
Pack of 10
Pack of 104
Pack of 20
Pack of 35
Pack of 60
Pack of 86
Product Code. 12820914
Edge
Store, pour or share your laboratory liquids using Round Glass Narrow Mouth Alpha Bottles. Available in clear or amber glass, these round bottles are an integral tool in the laboratory.
  • Narrow neck
  • Amber or clear glass
Specifications
Capacity (Metric) 100 mL
Color Amber
Material Soda Lime Glass
No. per Pack 86Pack
Quantity 86Pack
Mouth Narrow
Shape Round
Height (Metric) 105 mm
Product Type Alpha Bottle
Diameter (Metric) 46 mm