missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ RIN1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00€ - 508.00€
Specifications
| Antigen | RIN1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066880
|
Novus Biologicals™
NBP3-30812-25ug |
25 μg |
213.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067072
|
Novus Biologicals™
NBP3-30812-100ug |
100 μg |
508.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RIN1 Polyclonal antibody specifically detects RIN1 in Human samples. It is validated for Western BlotSpecifications
| RIN1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Ras and Rab interactor 1, ras inhibitor 1, Ras inhibitor JC99, ras inhibitor RIN1, Ras interaction/interference protein 1 | |
| His-tagged recombinant human RIN1 protein | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| Phosphate buffered saline | |
| 9610 | |
| IgG | |
| Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title